Monofunctional maleimide fluors, CyDye™
Supplier: Cytiva
PA13131 PA15130 PA15131 PA15136 PA15631 PA15636
PA13131EA 407 EUR
PA13131 PA15130 PA15131 PA15136 PA15631 PA15636
Monofunctional maleimide fluors, CyDye™
Protein Linking and Labeling Systems
Cy3 and Cy5 monofunctional maleimides are particularly suitable for the selective labelling of molecules containing free sulfhydryl groups, such as cysteine residues in proteins and peptides and oligonucleotides.
  • Particularly effective for labeling antibodies
  • Large range of bulk packs available for maximum convenience and economy
  • Example protocols included with each dye pack
Also useful in antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.