VWR®, Residual solvent standards according to ICH guidelines
Supplier: VWR Chemicals
88206.001EA 92.7 EUR
88206.001 88206.015 88207.001 88207.015 88208.001 88208.015 88209.001 88209.015
VWR®, Residual solvent standards according to ICH guidelines
Standards Organic Standards
The level of volatile organic solvents in pharmaceutical raw materials or finished product is defined and monitored in accordance with ICH guidelines. Residual solvents are classified according to their risk assessment classes. A selection of residual solvent standards is now available from Avantor.
  • Conforms to the latest Ph. Eur. edition
  • Conforms to ICH guidelines
  • Delivered with comprehensive Certificate of Analysis
  • Packed in 1 ml or 1,5 ml glass ampoules
Class I: Benzene, 1,2- dichloroethane, 1,1- dichloroethane, tetrachloromethane, 1,1,1- trichloroethane.
Class IIa: Chlorobenzene, cyclohexane, 1,2-dichloroethene (cis-/- trans), dichloromethane, dimethylformamide, ethylbenzene, n-hexane, methylcyclohexane, toluene, trichloroethene, m-xylene, o-xylene, p-xylene.
Class IIb: Acetonitrile, chloroform, 1,2-dimethoxyethane, N,N-dimethylacetamide, dioxane, 2-hexanone, methanol, nitromethane, pyridine, tetrahydronaphthalene.
Class IIc: Ethyleneglycol-monoethyl ether, ethyleneglycol, formamide, ethyleneglycol-monomethyl ether, 1-methyl-2-pyrrolidone, sulpholane.
Certifications: Conforms to the latest Ph. Eur. edition.
Conforms to ICH guidelines.
Delivery information: Supplied with a comprehensive Certificate of Analysis.
Packaging: Packed in 1 or 1,5 ml glass ampoules.