Di-tert-butyl iminodicarboxylate 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Di-(tert-butyl) imidodicarbonate
369660050 369660250 369661000
ACRO369660050EA 68.2 EUR
ACRO369660050 ACRO369660250 ACRO369661000
Di-tert-butyl iminodicarboxylate 97%
Di-tert-butyl iminodicarboxylate
Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043309
CAS Number: 51779-32-9
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %