γ-(N-Morpholino)propanesulphonic acid (MOPS) 1 mol/l aqueous solution, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
MQ200
M1442-100ML M1442-1L M1442-500ML
SIALM1442-100MLEA 102 EUR
SIALM1442-100ML SIALM1442-1L SIALM1442-500ML
γ-(N-Morpholino)propanesulphonic acid (MOPS) 1 mol/l aqueous solution, Sigma-Aldrich®
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Flash Pt: 110
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288