Ergocalciferol (vitamin D2, calciferol) 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (S,Z)-3-((E)-2-((1R,3aS,7aR)-1-((2R,5R,E)-5,6-Dimethylhept-3-en-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexanol

J62163.03 J62163.06
J62163.03EA 44.4 EUR
J62163.03 J62163.06
Ergocalciferol (vitamin D2, calciferol) 97%
Ergocalciferol (Vitamin D2, Calciferol)
Formula: C₂₈H₄₄O
MW: 396.66 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00166988
CAS Number: 50-14-6
EINECS: 200-014-9
UN: 2811
ADR: 6.1 II
Merck Index: 14,10018

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR