1H,1H,2H,2H-Perfluorooctyltriethoxysilane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;
M-Clarity™ quality level = MQ100
667420-25G 667420-5G
SIAL667420-25GEA 632 EUR
SIAL667420-25G SIAL667420-5G
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, Sigma-Aldrich®
1H,1H,2H,2H-Perfluorooctyltriethoxysilane
Formula: C₁₄H₁₉F₁₃O₃Si
MDL Number: MFCD00042333
CAS Number: 51851-37-7
EINECS: 257-473-3