1,4-Benzodioxane-6-boronic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,3-Dihydro-1,4-benzodioxine-6-boronic acid
M-Clarity™ quality level = MQ100
635995-1G 635995-5G
SIAL635995-1GEA 34.9 EUR
SIAL635995-1G SIAL635995-5G
1,4-Benzodioxane-6-boronic acid, Sigma-Aldrich®
1,4-Benzodioxane-6-boronic acid
Formula: C₈H₉BO₄
MDL Number: MFCD01009696
CAS Number: 164014-95-3