Guanine hydrochloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Amino-1H-purin-6(7H)-one hydrochloride

51030-10G 51030-50G
SIAL51030-10GEA 56.4 EUR
SIAL51030-10G SIAL51030-50G
Guanine hydrochloride, Sigma-Aldrich®
Guanine hydrochloride
Formula: C₅H₆ClN₅O
MDL Number: MFCD00213670
CAS Number: 635-39-2
EINECS: 211-235-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR