Trithiocyanuric acid 95%
Supplier: Thermo Fisher Scientific
Warning
140530050 140530250
ACRO140530050EA 24.3 EUR
ACRO140530050 ACRO140530250
Trithiocyanuric acid 95%
Trithiocyanuric acid
Formula: C₃H₃N₃S₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006052
CAS Number: 638-16-4
EINECS: 211-322-8
Order Now
Specification Test Results
Appearance Yellow Powder
Infrared spectrum Conforms
HPLC ≥94.0 %
GC ≥97.5 %