Trithiocyanuric acid 95%

Supplier: Thermo Fisher Scientific
Warning

A19080.06 A19080.14 A19080.22
A19080.06EA 23.9 EUR
A19080.06 A19080.14 A19080.22
Trithiocyanuric acid 95%
Trithiocyanuric acid

Purity: 95% CAS No.: 638-16-4 Molecular Formula: C3H3N3S3 Formula Weight: 177.27 Melting Point: 300° MDL No.: MFCD00006052

Formula: C₃H₃N₃S₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006052
CAS Number: 638-16-4
EINECS: 211-322-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR