Trithiocyanuric acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
T88595-25G
T88595-5G
SIALT88595-25GEA
160
EUR
SIALT88595-25G
SIALT88595-5G
Trithiocyanuric acid, Sigma-Aldrich®
Trithiocyanuric acid
Formula:
C₃H₃N₃S₃ |
MDL Number:
MFCD00006052 CAS Number: 638-16-4 EINECS: 211-322-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...