4'-(Methylsulphonyl)acetophenone ≥97%
Supplier: Thermo Fisher Scientific
Synonyms: 4'-(Methylsulphonyl)acetophenone
H54072.03 H54072.06
H54072.03EA 38.7 EUR
H54072.03 H54072.06
4'-(Methylsulphonyl)acetophenone ≥97%
4'-(Methylsulphonyl)acetophenone
Formula: C₉H₁₀O₃S
MW: 198.24 g/mol
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00025072
CAS Number: 10297-73-1
EINECS: 233-672-0