5-Chloro-2-benzoxazolinone ≥99%
Supplier: Thermo Scientific
|
Danger
|
B24507.18EA
48.8
EUR
B24507.18
5-Chloro-2-benzoxazolinone ≥99%
5-Chloro-2-benzoxazolinone
Formula:
C₇H₄ClNO₂ MW: 169,57 g/mol Melting Pt: 189…192 °C Storage Temperature: Ambient |
CAS Number:
95-25-0 Merck Index: 14,2194 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...