Europium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Europium(III) perchlorate
40572.NA 40572.NC
40572.NAEA 139 EUR
40572.NA 40572.NC
Europium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade
Europium (III) perchlorate hexahydrate
CAS No.: 13537-22-9 Molecular Formula: Eu(ClO4)3.6H2O Formula Weight: 558.40 (450.32anhy) Physical Form: Liquid Odour: Odourless UN No.: 3211 MDL No.: MFCD00150233
Formula: Cl₃EuO₁₂·6H₂O
MW: 558.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150233
CAS Number: 13537-22-9
EINECS: 236-907-5
UN: 3211
ADR: 5.1 II