Vincristine sulphate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

V8388-1MG V8388-5MG
SIALV8388-1MGEA 256 EUR
SIALV8388-1MG SIALV8388-5MG
Vincristine sulphate, Sigma-Aldrich®
Vincristine sulphate
Formula: C₄₆H₅₈N₄O₁₄S
MDL Number: MFCD00084729
CAS Number: 2068-78-2
EINECS: 218-190-0
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR