Trimethylamine 1 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
388371000 388378000
388371000.EA 86.7 EUR
388371000. 388378000.
Trimethylamine 1 M in THF, AcroSeal®
Trimethylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 2.87
Melting Pt: -117.1
Density: 0.85 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 2924
ADR: 3 II
Merck Index: 12,09841
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Strength 1 M Exact strength on the CoA