7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine

Supplier: Thermo Scientific

CC13210DA CC13210EA
MAYBCC13210DAEA 0 EUR
MAYBCC13210DA MAYBCC13210EA
7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine
7-Bromo-3,4-dihydro-2H-1,5-benzodioxepine
Formula: C₉H₉BrO₂
MDL Number: MFCD00508291
CAS Number: 147644-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR