Capillary columns, TraceGOLD™ TG-OCP
Supplier: Thermo Fisher Scientific
26078-5760 26077-1430 26077-5720 26078-1420
555-8972EA 892 EUR
555-8972 555-8811 555-8805 555-8799
Capillary columns, TraceGOLD™ TG-OCP
Chromatography Columns
Application-specific columns for organochlorine pesticides and herbicides.
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Fast analysis time giving full separation of chlorinated pesticides
  • Ideal for US EPA methods 8081, 608 and CLP