Manual detergent, alkaline, Sapphire
Supplier: TRISTEL SOLUTIONS
CRY301 CRY303
TRSTCRY301EA 150 EUR
TRSTCRY301 TRSTCRY303
Manual detergent, alkaline, Sapphire
Cleaning Agents Detergents and Surface Cleansers
A high foam, heavy duty, detergent for removal of organic soils, including fats, oils and silicone.
  • Alkaline detergent for organic soils
  • Dilution for use: between 1 to 5% v/v
Areas of use include, walls, floors, product contact areas and equipment such as vessels, filling lines and utensils. Sapphire is not soft metal safe, but can be used on a variety of surfaces including stainless steel.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
Delivery information: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
Packaging: Available in 20, 200 and 1000 litre pack sizes.