Glass density bottles acc. Gay-Lussac
Supplier: Witeg
WITG3.902.002EA 27.4 EUR
WITG3.902.002
Glass density bottles acc. Gay-Lussac
Specific Gravity Bottles
Made of borosilicate glass.
Pycnometer with with ground-in capillary stopper, precisely adjusted to the second place of decimal, acc. to ISO 3507.