Dodecacarbonyltriosmium ≥99%

Supplier: Thermo Fisher Scientific
Warning

13038.02 13038.04
13038.02EA 297 EUR
13038.02 13038.04
Dodecacarbonyltriosmium ≥99%
Dodecacarbonyltriosmium

Purity: 99% CAS No.: 15696-40-9 Molecular Formula: Os3(CO)12 Formula Weight: 906.73 Physical Form: Crystalline Melting Point: 224° UN No.: 3466 MDL No.: MFCD00011149

Formula: C₁₂O₁₂Os₃
Melting Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00011149
CAS Number: 15696-40-9
EINECS: 239-789-3
UN: 3288
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR