(±)-2-Piperazinecarboxylic acid dihydrochloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Piperazine-2-carboxylic acid dihydrochloride

M-Clarity™ quality level = MQ100

484903-25G 484903-5G
SIAL484903-25GEA 115 EUR
SIAL484903-25G SIAL484903-5G
(±)-2-Piperazinecarboxylic acid dihydrochloride, Sigma-Aldrich®
(±)-2-Piperazinecarboxylic acid dihydrochloride
Formula: C₅H₁₂Cl₂N₂O₂
MDL Number: MFCD00044803
CAS Number: 3022-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR