Diethyl aminomalonate hydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Diethyl 2-aminomalonate hydrochloride
M-Clarity™ quality level = MQ100
D87589-100G D87589-10G D87589-25G
SIALD87589-100GEA 249 EUR
SIALD87589-100G SIALD87589-10G SIALD87589-25G
Diethyl aminomalonate hydrochloride, Sigma-Aldrich®
Diethyl aminomalonate hydrochloride
Formula: C₇H₁₄ClNO₄
MDL Number: MFCD00012510
CAS Number: 13433-00-6
EINECS: 236-556-8