3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine

Supplier: Thermo Scientific

Synonyms: 5-Amino-3-(4-chlorophenyl)-1-methyl-1H-pyrazole

KM04038DA KM04038EA
MAYBKM04038DAEA 0 EUR
MAYBKM04038DA MAYBKM04038EA
3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
3-(4-Chlorophenyl)-1-methyl-1H-pyrazol-5-amine
Formula: C₁₀H₁₀ClN₃
MDL Number: MFCD00084872
CAS Number: 126417-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR