4-Fluorphenylmagnesium bromide 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Fluorophenylmagnesium bromide
M-Clarity™ quality level = MQ100
328820-100ML 328820-800ML
SIAL328820-100MLEA 67.1 EUR
SIAL328820-100ML SIAL328820-800ML
4-Fluorphenylmagnesium bromide 1 mol/l in THF, Sigma-Aldrich®
4-Fluorphenylmagnesium bromide
Formula: C₆H₄BrFMg
MDL Number: MFCD00009667
CAS Number: 352-13-6