4-Fluorphenylmagnesium bromide 2 mol/l in diethyl ether, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Fluorophenylmagnesium bromide
M-Clarity™ quality level = MQ100
245550-100ML 245550-800ML
SIAL245550-100MLEA 95.7 EUR
SIAL245550-100ML SIAL245550-800ML
4-Fluorphenylmagnesium bromide 2 mol/l in diethyl ether, Sigma-Aldrich®
4-Fluorphenylmagnesium bromide
Formula: C₆H₄BrFMg
MDL Number: MFCD00009667
CAS Number: 352-13-6