Lanthanum triacetate hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Lanthanum(III) acetate hydrate
M-Clarity™ quality level = MQ100
306339-100G
306339-500G
SIAL306339-100GEA
164
EUR
SIAL306339-100G
SIAL306339-500G
Lanthanum triacetate hydrate, Sigma-Aldrich®
Lanthanum triacetate hydrate
Formula:
C₆H₉LaO₆·H₂O MW: 334.053 g/mol |
MDL Number:
MFCD00150118 CAS Number: 100587-90-4 EINECS: 213-034-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...