(3-Benzyloxypropyl)triphenylphosphonium bromide ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: [3-(Benzyloxy)propyl]triphenylphosphonium Bromide
H55754.06 H55754.14
H55754.06EA 166 EUR
H55754.06 H55754.14
(3-Benzyloxypropyl)triphenylphosphonium bromide ≥97%
(3-Benzyloxypropyl)triphenylphosphonium bromide
Formula: C₂₈H₂₈BrOP
MW: 491.41 g/mol
Melting Pt: 153…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00191781
CAS Number: 54314-85-1