Magnesium ≥99.95% (metals basis), rods 6.35 mm (0.25 in) Ø x 25.4 mm (1.0 in) length,
Supplier: Thermo Fisher Scientific
47073.KF 47073.KJ 47073.KM
47073.KFEA 79.3 EUR
47073.KF 47073.KJ 47073.KM
Magnesium ≥99.95% (metals basis), rods 6.35 mm (0.25 in) Ø x 25.4 mm (1.0 in) length,
Magnesium
Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675