3-Bromo-5-formylpyridine ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-Bromopyridine-5-carboxaldehyde, 5-Bromopyridine-3-carboxaldehyde, 5-Bromonicotinaldehyde, 5-Bromo-3-pyridinecarboxaldehyde
H59224.03 H59224.06
H59224.03EA 52.7 EUR
H59224.03 H59224.06
3-Bromo-5-formylpyridine ≥97%
3-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD03265758
CAS Number: 113118-81-3