1,4-Dioxaspiro[4.5]decan-8-one, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
274879-100G 274879-25G 274879-5G
SIAL274879-100GEA 178 EUR
SIAL274879-100G SIAL274879-25G SIAL274879-5G
1,4-Dioxaspiro[4.5]decan-8-one, Sigma-Aldrich®
1,4-Dioxaspiro[4.5]decan-8-one
Formula: C₈H₁₂O₃
MDL Number: MFCD00010214
CAS Number: 4746-97-8