4-(Methylsulphonyl)phenylboronic acid 98+%
Supplier: Thermo Fisher Scientific
Warning
359510010 359510050
ACRO359510010EA 62.5 EUR
ACRO359510010 ACRO359510050
4-(Methylsulphonyl)phenylboronic acid 98+%
4-(Methylsulphonyl)phenylboronic acid
Formula: C₇H₉BO₄S
Melting Pt: 275…277 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01630820
CAS Number: 149104-88-1
Order Now
Specification Test Results
Appearance Off-white Powder
Infrared spectrum Conforms
HPLC ≥98.0 %