N-(2,4-Dichlorobenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide

Supplier: Thermo Scientific

RF04578.250 RF04578.1
MAYBRF04578.250EA 0 EUR
MAYBRF04578.250 MAYBRF04578.1
N-(2,4-Dichlorobenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide
N-(2,4-Dichlorobenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide
Formula: C₂₀H₁₀Cl₂F₁₃NO
MDL Number: MFCD02180605
CAS Number: 680213-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR