Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate ≥99%
Supplier: Thermo Fisher Scientific
Synonyms: Dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
B24008.22 B24008.36
B24008.22EA 49.2 EUR
B24008.22 B24008.36
Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate ≥99%
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate
Purity: 99+% CAS No.: 6289-46-9 Molecular Formula: C10H12O6 Formula Weight: 228.2 Melting Point: 152-156° MDL No.: MFCD00001607
Formula: C₁₀H₁₂O₆
MW: 228.2 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00001607
CAS Number: 6289-46-9
EINECS: 228-528-9