Industrial gloves, Harpon 321
Supplier: MAPA
34321000 34321166 34321007 34321008 34321009
112-5551EA 13.6 EUR
112-5551 112-5552 112-5553 112-5554 112-5555
Industrial gloves, Harpon 321
Gloves
These liquid-proof gloves are made of natural latex. They feature a cotton knit lining.
  • Reinforced grip: Ideal when gripping heavy, rough or slippery objects in highly aggressive environments
  • Suitable for long-term wear, due to the comfort of a textile support and good thermal insulation
  • Optimised service life: high resistance to abrasion
Suitable applications include: Fish-handling, oyster farming, work with frozen products.
Certifications: EN 388:2016 (3141X), EN ISO 374-1 Type B (KPT), EN 407 (X2XXXX); Cat. II