Chemical resistant gloves, Telsol 351
Supplier: MAPA
34351008 34351009 34351000
MAPA34351008EA 72.8 EUR
MAPA34351008 MAPA34351009 MAPA34351000
Chemical resistant gloves, Telsol 351
Gloves
PVC gloves with knitted textile support for improved comfort. The gloves provide protection against mechanical and low-risk chemical hazards.
  • Exceptional comfort and good resistance to cold, due to the seamless cotton-knit
  • Good resistance to oils, greases and hydrocarbons
  • Suitable for food contact, except fatty foods
  • Pebbled finish
Ideal for use in the pharmaceutical industry and for maintenance and cleaning tasks in wet environments (water, oil, greases, hydrocarbons).
Certifications: EN 388:2016 (4121X), EN ISO 374-1 Type A (KLMNPT), EN ISO 374-5; Cat. III; AQL <0,65.