Heat resistant gloves, TempDex 710
Supplier: MAPA
34710021 34710027 34710039
112-5102EA 9.1 EUR
112-5102 112-5100 MAPA34710039
Heat resistant gloves, TempDex 710
Gloves
These textured nitrile gloves with seamless textile support provide high dexterity and thermal protection.
  • Thin, anatomically shaped glove ensures dexterity
  • Excellent abrasion resistance: Longer lasting
  • Special nitrile grip coating facilitates work in moderately oily environments
  • Dot embossing for better insulation and better grip
  • Heat resistant up to a contact temperature of 125 °C
Ideal for handling composite materials after heat processing, handling moulds in the ceramic industry, handling hot steel after welding, as well as for handling hot piping. Also suitable for maintenance of pipes and heating equipment.
Certifications: EN 388:2016 (4111X), EN 407 (X1XXXX); Cat. II