Neutralising agent for the automated cleaning of glassware, neodisher® N
Supplier: Dr. Weigert
420130 420113 420126 420187 420133
148-3193EA 179 EUR
148-3193 148-3194 148-3195 141-0939 148-3192
Neutralising agent for the automated cleaning of glassware, neodisher® N
Cleaning Agents Detergents and Surface Cleansers
neodisher® N.
  • Acidic liquid concentrate for neutralisation of alkaline residues in the main automated cleaning cycle or for acidic pre-cleaning of laboratory glassware
  • Components: >30% phosphoric acid
  • Dosage: 1 to 2 ml/L for neutralisation; 3 to 4 ml/L for acidic pre-cleaning
Components: > 30% phosphoric acid
Dosage: 1 - 2 ml/l for neutralisation; 3 - 4 ml/l for acidic precleaning
Note: The materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.
Acidic liquid concentrate for neutralisation of alakaline residues in the main automted cleaning cyle or for acidic precleaning of laboratory glassware. Free of phosphates and surfactants
Components: Organic acids
Caution: The materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.