Cut resistant gloves, HyFlex® 11-435
Supplier: Ansell
11-435/6 11-435/7
112-4356EA 0 EUR
112-4356 112-4357
Cut resistant gloves, HyFlex® 11-435
Gloves
This knitted liner made of Dyneema®/glass fibre/Lycra® and nylon features a water-based PU coating on the palm.
  • Anti-static, breathable
  • Extreme resistance to cuts and burrs
  • Seamless plaited glove for very good fit, dexterity and flexibility
  • Water-based PU enhances the dexterity and comfort of the glove, as the PU doesn't penetrate inside it
The glove is ideal for the handling of glass and metal sheets, sharp-edged objects, pre- assembling, cutting of small dry or lightly oiled parts and maintenance tasks.
Certifications: EN 388:2016 (4X42C); Cat. II