Safety shoes, buckle fastening, Classic, 1500
Supplier: Abeba
1500/37 1500/38 1500/39 1500/40 1500/41 1500/42 1500/43 1500/44 1500/45 1500/36 1500/46
114-2355EA 85.6 EUR
114-2355 114-2356 114-2357 114-2358 114-2359 114-2360 114-2361 114-2362 114-2363 ABEB1500/36 ABEB1500/46
Safety shoes, buckle fastening, Classic, 1500
Shoes and Boots
Anti-static shoe made from grained leather and PU sole (resistant to animal fats, oil and fuel).
  • Comfortable shape
  • Microfibre lining
  • Non slip PU sole
  • Protective steel toecap (200 J)
  • Adjustable instep strap with buckle fastening
Suitable for use in kitchens and in food processing industries.
Colour: White
Certifications: EN ISO 20345; S2, SRC