Hand cleanser dispensers, Eurospender, 1 Plus
Supplier: BODE CHEMIE
980478 980479 981098 981099 981125 9812510 9812550 9812530
115-4031EA 98.8 EUR
115-4031 115-4032 BODE981098 BODE981099 BODE981125 BODL9812510 BODL9812550 BODL9812530
Hand cleanser dispensers, Eurospender, 1 Plus
Soap Dispensers and Lotion Soap Dispensers
The Eurospender 1 Plus are versatile and user-friendly wall-mounting dispensers. The pump can be quickly exchanged through front removal.
  • Robust and reliable metal dispensers which meet high hygiene standards
  • Dosing amount adjustable to approx. 0,75 to 1,5 ml per actuation
  • Dispensing carried out via the operating lever
Delivery information: Including mounting parts, instruction sheet and manual. Accessories and spare parts are available on request. Please contact your local Avantor customer service centre for further information.