UHPLC columns, Pinnacle® DB Silica (USP L3)
Supplier: Restek
9410512-700 9410513-700 9410515-700 9410522-700 9410523-700 9410525-700 9410532-700 9410533-700 9410535-700 9410552-700 9410553-700 9410555-700 9410562-700 9410563-700 9410565-700 9410572-700 9410573-700 9410575-700 9410512 9410515 9410522 9410525 9410532 9410535 9410552 9410555 9410562 9410565 9410572 9410575 941050210 941050212 9410212 9410232 9410252 9410312 9410312-700 9410313-700 9410315 9410315-700 941031E 9410332 9410332-700 9410333-700 9410335 9410335-700 941033E 9410352 9410352-700 9410353-700 9410355 9410355-700 941035E 9410362 9410362-700 9410363-700 9410365 9410365-700 941036E 941051E 941052E 941053E 941055E 941056E 941057E
REST9410512-700EA 603 EUR
REST9410512-700 REST9410513-700 REST9410515-700 REST9410522-700 REST9410523-700 REST9410525-700 REST9410532-700 REST9410533-700 REST9410535-700 REST9410552-700 REST9410553-700 REST9410555-700 REST9410562-700 REST9410563-700 REST9410565-700 REST9410572-700 REST9410573-700 REST9410575-700 REST9410512 REST9410515 REST9410522 REST9410525 REST9410532 REST9410535 REST9410552 REST9410555 REST9410562 REST9410565 REST9410572 REST9410575 REST941050210 REST941050212 554-1552 554-1553 554-1554 REST9410312 REST9410312-700 REST9410313-700 REST9410315 REST9410315-700 REST941031E REST9410332 REST9410332-700 REST9410333-700 REST9410335 REST9410335-700 REST941033E REST9410352 REST9410352-700 REST9410353-700 REST9410355 REST9410355-700 REST941035E REST9410362 REST9410362-700 REST9410363-700 REST9410365 REST9410365-700 REST941036E REST941051E REST941052E REST941053E REST941055E REST941056E REST941057E
UHPLC columns, Pinnacle® DB Silica (USP L3)
Chromatography Columns
Highly base-deactivated spherical silica manufactured by Restek. Useful for normal phase separations. Replaces Hypersil® BDS and Pinnacle® Amine.