Safety ankle boots, slip-on, Steelite™ MICROFIBRE FW83
Supplier: Portwest
FW83WHR35 FW83WHR36 FW83WHR37 FW83WHR38 FW83WHR39 FW83WHR41 FW83WHR42 FW83WHR43 FW83WHR44 FW83WHR45 FW83WHR46 FW83WHR47 FW83WHR48
PRTWFW83WHR35EA 43.5 EUR
PRTWFW83WHR35 PRTWFW83WHR36 PRTWFW83WHR37 PRTWFW83WHR38 PRTWFW83WHR39 PRTWFW83WHR41 PRTWFW83WHR42 PRTWFW83WHR43 PRTWFW83WHR44 PRTWFW83WHR45 PRTWFW83WHR46 PRTWFW83WHR47 PRTWFW83WHR48
Safety ankle boots, slip-on, Steelite™ MICROFIBRE FW83
Shoes and Boots
Anti-static lab shoe manufactured from high quality water resistant microfibre and a monodensity PU sole resistant to oil and heat (200 °C). Ideal for use in cleanrooms, medical or food industry.
  • Padded collars provide all day long comfort
  • Shock absorber in heel
  • Machine washable at 30 °C
  • Self cleaning sole to prevent cross contamination
  • Protective toecap made from steel (200 J)
Colour: White
EN ISO 20345:2011; S2, SRC