Safety shoes, lace-up, Steelite™ Executive Brogue FW46
Supplier: Portwest
FW46BKR39 FW46BKR47 FW46BKR46 FW46BKR45 FW46BKR44 FW46BKR43 FW46BKR42 FW46BKR40 FW46BKR41
PRTWFW46BKR39EA 85.1 EUR
PRTWFW46BKR39 PRTWFW46BKR47 PRTWFW46BKR46 PRTWFW46BKR45 PRTWFW46BKR44 PRTWFW46BKR43 PRTWFW46BKR42 PRTWFW46BKR40 PRTWFW46BKR41
Safety shoes, lace-up, Steelite™ Executive Brogue FW46
Shoes and Boots
These laced executive brogue office/service shoes are manufactured from Brazilian full grain leather plus an oil resistant, dual-density PU/PU outer sole.
  • Special moisture management fabric for extra comfort
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certifications: EN ISO 20345:2011; S1P, SRC.