Flame-retardant overalls with bib and braces, high visibility, Brisbane 6133
Supplier: SIOEN
6133/278/1XL 6133/278/2XL 6133/278/3XL 6133/278/L 6133/278/M 6133/278/S 6133/279/1XL 6133/279/2XL 6133/279/3XL 6133/279/L 6133/279/M 6133/279/S
SIOE6133/278/1XLEA 177 EUR
SIOE6133/278/1XL SIOE6133/278/2XL SIOE6133/278/3XL SIOE6133/278/L SIOE6133/278/M SIOE6133/278/S SIOE6133/279/1XL SIOE6133/279/2XL SIOE6133/279/3XL SIOE6133/279/L SIOE6133/279/M SIOE6133/279/S
Flame-retardant overalls with bib and braces, high visibility, Brisbane 6133
Workwear
These water- and wind-proof highly breathable bib & brace overalls are made of 100% polyester fabric with 100% FR PU coating + AST. They also feature a fixed flame retardant cotton lining.
  • Water repellent outer fabric, moisture attracting coating on the inside
  • Braces with buckle, front bib
  • Leg narrowing below by touch and close fastening
  • Zip closure under flap with touch and close fastening at leg end
  • Taped seams
  • Flame retardant reflective tape (50 mm)
  • Anti-static
Weight: 250 g/m²
Certifications: EN 531 (A, B1, C1), EN 533 (Index 3), EN 1149-5, EN 13034 (type PB [6]), EN 343 (3-3), EN 471 (2-2)