Overalls with bib and braces, Oak Chainsaw CH12
Supplier: Portwest
CH12BKR1XL CH12BKR2XL CH12BKR3XL CH12BKRL CH12BKRM CH12BKRS
PRTWCH12BKR1XLEA 151 EUR
PRTWCH12BKR1XL PRTWCH12BKR2XL PRTWCH12BKR3XL PRTWCH12BKRL PRTWCH12BKRM PRTWCH12BKRS
Overalls with bib and braces, Oak Chainsaw CH12
Workwear
These trousers have 9 layers of protection to the front of each leg and are mainly intended to be used for forestry work by professional lumberjacks who work with both feet on the ground. Made of 65% polyester and 35% cotton on the outside and polyester/PP fabric on the inside, the trousers are sweat wicking, light and flexible.
  • Excellent breathable and water-resistant properties
  • Stretch fabric in key areas for greater ease of movement
  • Brace lock and brace length adjuster
  • Front zipper opening, elasticated waist and extended back
  • Two inset pockets
Weight (outer fabric): 245 g/m²
Weight (inner fabric): 120 g/m²
Certifications: EN 381-5:1995, class 1, type A, 20 M/S