Chemiluminescent detection reagents, Luminata™ Western HRP substrates
Supplier: Merck
WBLUF0500 WBLUR0500 WBLUF0100 WBLUC0100 WBLUC0500 WBLUR0100
MILFWBLUF0500EA 521 EUR
MILFWBLUF0500 MILFWBLUR0500 MILFWBLUF0100 MILFWBLUC0100 MILFWBLUC0500 MILFWBLUR0100
Chemiluminescent detection reagents, Luminata™ Western HRP substrates
Electrophoresis Reagents Western Blotting Reagents
Luminata™ Western HRP substrates are a family of premixed, ready to use chemiluminescent reagents for the detection of HRP-based Westerns.
  • Premixed for convenience and to avoid cross contamination of the reagents
  • Consistent results with less pipetting error
  • Stable at 4 °C or room temperature so it can be stored on the bench
  • Compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage conditions Stable for 1 year at room temperature from date of shipment Stable for 1 year at 4 °C from date of shipment
Storage temperature 18...24 °C 2...8 °C