AccuStart™ II PCR ToughMix®
Supplier: Quantabio
95142-04K 95142-800 95142-100
733-2259EA 2270 EUR
733-2259 733-2261 QUNT95142-100
AccuStart™ II PCR ToughMix®
Nucleic Acid Reagents End-point PCR Enzymes and Kits
AccuStart™ II PCR ToughMix® is a 2X concentrated ready to use reaction cocktail for PCR amplification of DNA templates that overcomes many known inhibitors of PCR often present in crude samples extracted from environmental specimens, plant tissues, or animal tissues. It contains all components, except primers and template.
Kit components:
AccuStart II GelTrack™ PCR ToughMix®: 2X mix containing optimised concentrations of MgCl₂, dNTPs, reaction buffer, hot start DNA polymerase, stabilisers and gel loading dyes.
GelTrack™ Loading Dye: 50X concentrated mixture of RT-PCR compatible, blue and yellow electrophoresis tracking dyes.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more