ESD occupational shoes, clogs, Tiefbett Leder, Ingolstadt, 11312
Supplier: SCHURR SCHUHVERTRIEB
1.1312.01.41 1.1312.01.43 1.1312.01.42 1.1312.01.44 1.1312.01.45 1.1312.01.46 1.1312.01.47 1.1312.01.48 1.1312.04.41 1.1312.04.42 1.1312.04.43 1.1312.04.44 1.1312.04.45 1.1312.04.46 1.1312.04.47 1.1312.04.48 1.1412.01.1435 1.1412.01.1436 1.1412.01.1437 1.1412.01.1438 1.1412.01.1439 1.1412.01.1440
SCRR1.1312.01.41EA 60.3 EUR
SCRR1.1312.01.41 SCRR1.1312.01.43 SCRR1.1312.01.42 SCRR1.1312.01.44 SCRR1.1312.01.45 SCRR1.1312.01.46 SCRR1.1312.01.47 SCRR1.1312.01.48 SCRR1.1312.04.41 SCRR1.1312.04.42 SCRR1.1312.04.43 SCRR1.1312.04.44 SCRR1.1312.04.45 SCRR1.1312.04.46 SCRR1.1312.04.47 SCRR1.1312.04.48 SCRR1.1412.01.1435 SCRR1.1412.01.1436 SCRR1.1412.01.1437 SCRR1.1412.01.1438 SCRR1.1412.01.1439 SCRR1.1412.01.1440
ESD occupational shoes, clogs, Tiefbett Leder, Ingolstadt, 11312
Shoes and Boots
Deeply moulded occupational shoes made from full grain leather and an anti-slip and anti-static 'A4' nitrile rubber sole; with adjustable heel strap.
  • Anatomically shaped stabilising integrated insole made from cork/latex
  • Insole features cushioned heel, ankle support and toe bar
  • Shock absorber heel
For ESD and kitchen use.
Colour: White or blue
Width: 9
Weight: 265 g
Certifications: EN 61340 (ESD); EN ISO 20347:2012; OB, SRC