ESD safety shoes, clogs, SRC, Bamberg, 21381
Supplier: SCHURR SCHUHVERTRIEB
2.1481.01.37 2.1481.02.37 2.1381.01.43 2.1381.02.43 2.1381.01.44 2.1381.01.48 2.1481.02.40 2.1381.02.44 2.1381.01.42 2.1481.01.36 2.1481.01.38 2.1481.01.39 2.1481.01.40 2.1381.01.45 2.1381.01.41 2.1381.02.42 2.1381.02.46 2.1381.01.46 2.1381.01.47 2.1481.02.38 2.1381.02.41 2.1381.02.45 2.1381.02.47 2.1381.02.48 2.1481.02.36 2.1481.02.39
SCRR2.1481.01.37EA 90.3 EUR
SCRR2.1481.01.37 SCRR2.1481.02.37 SCRR2.1381.01.43 SCRR2.1381.02.43 SCRR2.1381.01.44 SCRR2.1381.01.48 SCRR2.1481.02.40 SCRR2.1381.02.44 SCRR2.1381.01.42 SCRR2.1481.01.36 SCRR2.1481.01.38 SCRR2.1481.01.39 SCRR2.1481.01.40 SCRR2.1381.01.45 SCRR2.1381.01.41 SCRR2.1381.02.42 SCRR2.1381.02.46 SCRR2.1381.01.46 SCRR2.1381.01.47 SCRR2.1481.02.38 SCRR2.1381.02.41 SCRR2.1381.02.45 SCRR2.1381.02.47 SCRR2.1381.02.48 SCRR2.1481.02.36 SCRR2.1481.02.39
ESD safety shoes, clogs, SRC, Bamberg, 21381
Shoes and Boots
ESD safety shoe made from microfibre Cordura®, DRYtech® lining with Cambrelle® mesh insert and an anti-slip and anti-static 'A4' nitrile rubber sole; with adjustable heel strap.
  • Antibacterial and breathable DRYtech® textile lining
  • Removable insole made from PUR-foam
  • Orthopedic insoles possible
  • Triple shock absorber at heel and ball of the foot relieves joints
  • Protective steel toe cap (200 J)
For ESD and kitchen use.
Colour: White or black
Width: 9
Weight: 505 g
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; SB, SRC